Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205225.2 | NP_990556.1 | 5 - 6 | 673 | 59 | |
GenBank mRNA | AF208228.1 | - | 4 - 5 | 651 | 59 | |
X81968.1 | - | 674 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |