Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205353.2 | NP_990684.1 | 7 - 8 | 789 | 92 | |
XM_025142488.1 | XP_024998256.1 | 7 - 8 | 1610 | 92 | ||
GenBank mRNA | X17700.1 | - | 781 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |