Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205404.1 | NP_990735.1 | 2 - 3 | 932 | 76 | |
GenBank mRNA | L35555.1 | - | 2 - 3 | 932 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |