Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205414.1 | NP_990745.1 | 5 - 6 | 1041 | 67 | |
XM_015277257.2 | XP_015132743.1 | 5 - 6 | 1337 | 67 | ||
XM_015277259.2 | XP_015132745.1 | 5 - 6 | 1343 | 67 | ||
XM_025144692.1 | XP_025000460.1 | 5 - 6 | 1330 | 67 | ||
GenBank mRNA | L35567.1 | - | 5 - 6 | 1041 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |