Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205421.2 | NP_990752.2 | 3 - 4 | 812 | 76 | |
XM_015296342.2 | XP_015151828.2 | 4 - 5 | 1157 | 76 | ||
XM_015296343.2 | XP_015151829.2 | 5 - 6 | 1045 | 76 | ||
GenBank mRNA | U11888.1 | - | 3 - 4 | 812 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |