Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205424.1 | NP_990755.1 | 12 - 13 | 1829 | 72 | |
GenBank mRNA | CR386644.1 | - | 9 - 10 | 1352 | 72 | |
L12144.1 | - | 12 - 13 | 1829 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |