Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006360.1 | NP_001006360.1 | 7 - 8 | 1008 | 115 | |
XM_025147197.1 | XP_025002965.1 | 8 - 9 | 1306 | 115 | ||
GenBank mRNA | AJ719598.1 | - | 7 - 8 | 1008 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |