Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278145.1 | NP_001265074.1 | 6 - 7 | 1039 | 136 | |
NR_103454.1 | 5 - 6 | 884 | 136 | |||
GenBank mRNA | AJ719981.1 | - | 5 - 6 | 883 | 136 | |
EU419873.1 | - | 6 - 7 | 941 | 136 | ||
HQ896422.1 | - | 6 - 7 | 941 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |