Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006375.1 | NP_001006375.1 | 2 - 3 | 282 | 134 | |
XM_015275890.2 | XP_015131376.1 | 6 - 7 | 2241 | 134 | ||
XM_015275891.2 | XP_015131377.1 | 6 - 7 | 2241 | 134 | ||
GenBank mRNA | AJ719521.1 | - | 2 - 3 | 282 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |