Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006575.2 | NP_001006575.1 | 3 - 4 | 903 | 112 | |
GenBank mRNA | AJ719604.1 | - | 3 - 4 | 903 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |