Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008676.2 | NP_001008676.1 | 6 - 7 | 996 | 84 | |
GenBank mRNA | AY820753.1 | - | 6 - 7 | 836 | 84 | |
AY957508.1 | - | 6 - 7 | 996 | 84 | ||
BX931414.2 | - | 5 - 6 | 771 | 84 | ||
CR390008.1 | - | 1 - 2 | 286 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |