Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012551.1 | NP_001012569.1 | 1 - 2 | 110 | 83 | |
GenBank mRNA | AJ851582.1 | - | 1 - 2 | 110 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |