Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012698.1 | NP_001012716.1 | 1 - 2 | 91 | 98 | |
GenBank mRNA | AJ851490.1 | - | 1 - 2 | 91 | 98 | |
CR406940.1 | - | 1 - 2 | 71 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |