Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012771.2 | NP_001012789.1 | 5 - 6 | 480 | 71 | |
XM_015292113.2 | XP_015147599.1 | 5 - 6 | 481 | 71 | ||
GenBank mRNA | AJ720180.1 | - | 5 - 6 | 481 | 71 | |
CR387918.1 | - | 3 - 4 | 280 | 71 | ||
CR523714.1 | - | 2 - 3 | 205 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |