Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012819.1 | NP_001012837.1 | 2 - 3 | 878 | 66 | |
XM_015295303.2 | XP_015150789.2 | 2 - 3 | 1538 | 66 | ||
XM_015295313.2 | XP_015150799.2 | 2 - 3 | 1538 | 66 | ||
XM_015295319.2 | XP_015150805.2 | 2 - 3 | 1538 | 66 | ||
XM_015295324.2 | XP_015150810.2 | 2 - 3 | 1538 | 66 | ||
XM_015295341.2 | XP_015150827.2 | 2 - 3 | 1538 | 66 | ||
XM_025149234.1 | XP_025005002.1 | 2 - 3 | 924 | 66 | ||
GenBank mRNA | AJ851407.1 | - | 2 - 3 | 878 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |