Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012820.1 | NP_001012838.1 | 3 - 4 | 510 | 132 | |
XM_015295972.2 | XP_015151458.1 | 3 - 4 | 630 | 132 | ||
XM_015295983.2 | XP_015151469.1 | 2 - 3 | 763 | 132 | ||
XM_015295985.2 | XP_015151471.1 | 2 - 3 | 753 | 132 | ||
GenBank mRNA | AJ720437.1 | - | 3 - 4 | 510 | 132 | |
CR732891.1 | - | 2 - 3 | 694 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |