Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012833.1 | NP_001012851.1 | 2 - 3 | 406 | 83 | |
XM_015296384.2 | XP_015151870.2 | 2 - 3 | 57 | 83 | ||
GenBank mRNA | AJ719437.1 | - | 2 - 3 | 398 | 83 | |
AJ851651.1 | - | 2 - 3 | 406 | 83 | ||
BX933801.1 | - | 2 - 3 | 144 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |