Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012866.1 | NP_001012884.1 | 6 - 7 | 1670 | 89 | |
GenBank mRNA | AJ851615.1 | - | 6 - 7 | 1670 | 89 | |
CR387415.1 | - | 5 - 6 | 568 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |