Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012874.2 | NP_001012892.2 | 3 - 4 | 445 | 116 | |
XM_015283646.2 | XP_015139132.1 | 2 - 3 | 200 | 116 | ||
XM_015283647.2 | XP_015139133.1 | 3 - 4 | 385 | 116 | ||
XM_015283648.2 | XP_015139134.1 | 2 - 3 | 288 | 116 | ||
GenBank mRNA | BX930248.2 | - | 2 - 3 | 180 | 116 | |
BX931514.1 | - | 3 - 4 | 400 | 116 | ||
CR385312.1 | - | 2 - 3 | 191 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |