Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012874.2 | NP_001012892.2 | 8 - 9 | 994 | 86 | |
XM_015283646.2 | XP_015139132.1 | 7 - 8 | 749 | 86 | ||
XM_015283647.2 | XP_015139133.1 | 8 - 9 | 934 | 86 | ||
XM_015283648.2 | XP_015139134.1 | 7 - 8 | 837 | 86 | ||
GenBank mRNA | BX930248.2 | - | 7 - 8 | 729 | 86 | |
BX931514.1 | - | 8 - 9 | 949 | 86 | ||
BX935292.2 | - | 7 - 8 | 739 | 86 | ||
CR385312.1 | - | 7 - 8 | 740 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |