Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012910.1 | NP_001012928.1 | 3 - 4 | 688 | 87 | |
XM_015288647.2 | XP_015144133.1 | 3 - 4 | 823 | 87 | ||
XM_015288648.2 | XP_015144134.1 | 3 - 4 | 710 | 87 | ||
XM_025151610.1 | XP_025007378.1 | 3 - 4 | 753 | 87 | ||
GenBank mRNA | AJ719833.1 | - | 3 - 4 | 688 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |