Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012936.2 | NP_001012954.2 | 1 - 2 | 361 | 54 | |
GenBank mRNA | AJ720998.1 | - | 1 - 2 | 330 | 54 | |
BU383289.1 | - | 1 - 2 | 73 | 54 | ||
CD217943.1 | - | 1 - 2 | 361 | 54 | ||
CR388843.1 | - | 1 - 2 | 169 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |