Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024833.2 | NP_001020004.2 | 4 - 5 | 298 | 140 | |
GenBank mRNA | AY954510.1 | - | 2 - 3 | 212 | 140 | |
CR352860.1 | - | 4 - 5 | 297 | 140 | ||
EU686392.1 | - | 4 - 5 | 296 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |