Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024833.2 | NP_001020004.2 | 5 - 6 | 404 | 115 | |
GenBank mRNA | AY954510.1 | - | 3 - 4 | 318 | 115 | |
CR352860.1 | - | 5 - 6 | 403 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |