Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030008.1 | NP_001025179.1 | 3 - 4 | 369 | 139 | |
XM_015276903.2 | XP_015132389.1 | 3 - 4 | 606 | 139 | ||
GenBank mRNA | BX931554.1 | - | 3 - 4 | 357 | 139 | |
CD762344.1 | - | 3 - 4 | 395 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |