Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030627.1 | NP_001025798.1 | 6 - 7 | 917 | 89 | |
GenBank mRNA | AJ720764.1 | - | 6 - 7 | 917 | 89 | |
CR524108.1 | - | 4 - 5 | 750 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |