Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030868.2 | NP_001026039.2 | 4 - 5 | 340 | 91 | |
XM_015296408.2 | XP_015151894.1 | 4 - 5 | 387 | 91 | ||
XM_015296409.2 | XP_015151895.1 | 4 - 5 | 365 | 91 | ||
XM_025142256.1 | XP_024998024.1 | 4 - 5 | 362 | 91 | ||
GenBank mRNA | AJ720526.1 | - | 340 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |