Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030868.2 | NP_001026039.2 | 7 - 8 | 584 | 59 | |
XM_015296408.2 | XP_015151894.1 | 7 - 8 | 631 | 59 | ||
XM_015296409.2 | XP_015151895.1 | 7 - 8 | 609 | 59 | ||
XM_025142256.1 | XP_024998024.1 | 7 - 8 | 606 | 59 | ||
GenBank mRNA | AJ720526.1 | - | 584 | 59 | ||
BU426351.1 | - | 2 - 3 | 143 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |