Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030874.1 | NP_001026045.1 | 4 - 5 | 610 | 107 | |
GenBank mRNA | AJ719323.1 | - | 4 - 5 | 610 | 107 | |
MH492023.1 | - | 4 - 5 | 469 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |