Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030885.2 | NP_001026056.2 | 3 - 4 | 439 | 92 | |
GenBank mRNA | AJ719734.1 | - | 3 - 4 | 437 | 92 | |
BU299154.1 | - | 3 - 4 | 439 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |