Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031067.1 | NP_001026238.1 | 3 - 4 | 256 | 69 | |
XM_015286169.2 | XP_015141655.2 | 3 - 4 | 264 | 69 | ||
XM_015286170.2 | XP_015141656.1 | 3 - 4 | 339 | 69 | ||
GenBank mRNA | AJ851475.1 | - | 3 - 4 | 256 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |