Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031067.1 | NP_001026238.1 | 4 - 5 | 462 | 84 | |
XM_015286169.2 | XP_015141655.2 | 4 - 5 | 470 | 84 | ||
GenBank mRNA | AJ851475.1 | - | 4 - 5 | 462 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |