Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031220.1 | NP_001026391.1 | 5 - 6 | 857 | 111 | |
XM_015288120.2 | XP_015143606.1 | 4 - 5 | 915 | 111 | ||
XM_015288128.2 | XP_015143614.1 | 12 - 13 | 5174 | 111 | ||
GenBank mRNA | AJ720565.1 | - | 5 - 6 | 857 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |