Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001337.1 | NP_001001337.1 | 9 - 10 | 1016 | 66 | |
NM_001001340.2 | NP_001001340.1 | 7 - 8 | 686 | 66 | ||
NM_001001341.1 | NP_001001341.1 | 7 - 8 | 803 | 66 | ||
NM_001306152.2 | NP_001293081.1 | 9 - 10 | 977 | 66 | ||
NM_204771.1 | NP_990102.1 | 8 - 9 | 938 | 66 | ||
GenBank mRNA | BU320294.1 | - | 7 - 8 | 616 | 66 | |
U04834.1 | - | 8 - 9 | 901 | 66 | ||
U06848.1 | - | 9 - 10 | 979 | 66 | ||
U06849.1 | - | 9 - 10 | 940 | 66 | ||
U06850.1 | - | 8 - 9 | 910 | 66 | ||
U06851.1 | - | 7 - 8 | 650 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |