Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001528.2 | NP_001001528.2 | 4 - 5 | 739 | 86 | |
GenBank mRNA | DN855767.1 | - | 1 - 2 | 68 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |