Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001529.1 | NP_001001529.1 | 1 - 2 | 438 | 63 | |
GenBank mRNA | AY463528.1 | - | 1 - 2 | 438 | 63 | |
BX931522.1 | - | 1 - 2 | 461 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |