Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001604.1 | NP_001001604.1 | 4 - 5 | 545 | 74 | |
XM_015292082.2 | XP_015147568.1 | 3 - 4 | 586 | 74 | ||
GenBank mRNA | AJ699419.1 | - | 4 - 5 | 545 | 74 | |
CR406140.1 | - | 3 - 4 | 540 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |