Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001741.1 | NP_001001741.1 | 2 - 3 | 314 | 81 | |
XM_015281332.2 | XP_015136818.1 | 2 - 3 | 81 | |||
GenBank mRNA | AJ251273.1 | - | 2 - 3 | 314 | 81 | |
KJ465915.1 | - | 1 - 2 | 267 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |