Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001879.1 | NP_001001879.1 | 3 - 4 | 479 | 112 | |
GenBank mRNA | BX932720.1 | - | 3 - 4 | 484 | 112 | |
CR353819.1 | - | 3 - 4 | 549 | 112 | ||
CR353824.1 | - | 3 - 4 | 479 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |