Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004369.1 | NP_001004369.1 | 5 - 6 | 656 | 78 | |
XM_015278892.2 | XP_015134378.1 | 6 - 7 | 797 | 78 | ||
XM_015278893.2 | XP_015134379.1 | 5 - 6 | 883 | 78 | ||
GenBank mRNA | X62531.1 | - | 656 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |