Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004374.2 | NP_001004374.1 | 1 - 2 | 133 | 76 | |
GenBank mRNA | CK987208.1 | - | 1 - 2 | 133 | 76 | |
CR387670.1 | - | 2 - 3 | 494 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |