Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004375.2 | NP_001004375.1 | 1 - 2 | 119 | 96 | |
GenBank mRNA | BX274578.4 | - | 1 - 2 | 125 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |