Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004376.3 | NP_001004376.1 | 1 - 2 | 122 | 100 | |
GenBank mRNA | BX261763.4 | - | 1 - 2 | 122 | 100 | |
V00380.1 | - | 111 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |