Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004379.1 | NP_001004379.1 | 6 - 7 | 697 | 83 | |
GenBank mRNA | CR385365.1 | - | 6 - 7 | 702 | 83 | |
X62640.1 | - | 697 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |