Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204153.1 | NP_989484.1 | 3 - 4 | 385 | 149 | |
XM_015276098.2 | XP_015131584.1 | 3 - 4 | 765 | 149 | ||
GenBank mRNA | AF000631.1 | - | 3 - 4 | 345 | 149 | |
AF017645.1 | - | 3 - 4 | 369 | 149 | ||
AF033563.1 | - | 3 - 4 | 385 | 149 | ||
AF483599.1 | - | 3 - 4 | 342 | 149 | ||
AF483600.1 | - | 3 - 4 | 342 | 149 | ||
AF502412.1 | - | 3 - 4 | 342 | 149 | ||
AY029588.1 | - | 3 - 4 | 337 | 149 | ||
AY370777.1 | - | 3 - 4 | 322 | 149 | ||
AY370778.1 | - | 3 - 4 | 322 | 149 | ||
AY510091.1 | - | 3 - 4 | 330 | 149 | ||
AY766162.1 | - | 3 - 4 | 325 | 149 | ||
GU119890.1 | - | 3 - 4 | 325 | 149 | ||
HQ008780.1 | - | 3 - 4 | 342 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |