Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204231.2 | NP_989562.2 | 2 - 3 | 646 | 95 | |
GenBank mRNA | AB088686.1 | - | 2 - 3 | 603 | 95 | |
AJ719767.1 | - | 2 - 3 | 646 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |