Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204270.4 | NP_989601.2 | 4 - 5 | 588 | 97 | |
GenBank mRNA | AB031025.1 | - | 4 - 5 | 549 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |