Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204849.1 | NP_990180.1 | 1 - 2 | 94 | 65 | |
GenBank mRNA | AF131224.1 | - | 1 - 2 | 94 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |