Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204958.1 | NP_990289.1 | 8 - 9 | 1047 | 138 | |
GenBank mRNA | U60187.1 | - | 8 - 9 | 1047 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |