Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205026.1 | NP_990357.1 | 15 - 16 | 1321 | 64 | |
XM_025142176.1 | XP_024997944.1 | 15 - 16 | 1345 | 64 | ||
GenBank mRNA | AB219157.1 | - | 14 - 15 | 1272 | 64 | |
AB219158.1 | - | 14 - 15 | 1272 | 64 | ||
AB219159.1 | - | 14 - 15 | 1272 | 64 | ||
AB219160.1 | - | 14 - 15 | 1272 | 64 | ||
AF029841.1 | - | 15 - 16 | 1321 | 64 | ||
HG007958.1 | - | 15 - 16 | 1351 | 64 | ||
LC144559.1 | - | 14 - 15 | 1272 | 64 | ||
LC144566.1 | - | 14 - 15 | 1272 | 64 | ||
LC144568.1 | - | 14 - 15 | 1272 | 64 | ||
LC144569.1 | - | 14 - 15 | 1272 | 64 | ||
LC144570.1 | - | 14 - 15 | 1272 | 64 | ||
LC144571.1 | - | 14 - 15 | 1272 | 64 | ||
LC144573.1 | - | 14 - 15 | 1272 | 64 | ||
LC144575.1 | - | 14 - 15 | 1272 | 64 | ||
LC144576.1 | - | 14 - 15 | 1272 | 64 | ||
LC144577.1 | - | 14 - 15 | 1272 | 64 | ||
LC144578.1 | - | 14 - 15 | 1272 | 64 | ||
LC144579.1 | - | 14 - 15 | 1272 | 64 | ||
LC144581.1 | - | 14 - 15 | 1272 | 64 | ||
LC144585.1 | - | 14 - 15 | 1272 | 64 | ||
LC144586.1 | - | 14 - 15 | 1272 | 64 | ||
LC144587.1 | - | 14 - 15 | 1272 | 64 | ||
LC144588.1 | - | 14 - 15 | 1272 | 64 | ||
LC144590.1 | - | 14 - 15 | 1272 | 64 | ||
LC144591.1 | - | 14 - 15 | 1272 | 64 | ||
LC144602.1 | - | 14 - 15 | 1272 | 64 | ||
LC144603.1 | - | 14 - 15 | 1272 | 64 | ||
LC144604.1 | - | 14 - 15 | 1272 | 64 | ||
LC144605.1 | - | 14 - 15 | 1272 | 64 | ||
LC144606.1 | - | 14 - 15 | 1272 | 64 | ||
LC144608.1 | - | 14 - 15 | 1272 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |